missing translation for 'onlineSavingsMsg'
Learn More

Theobromine, MedChemExpress

Product Code. 16488996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488996 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488996 Supplier Medchem Express Supplier No. HYN0138
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Theobromine is a methylxanthine found in cacao beans which can inhibit adenosine receptor A1 (AR1) signaling.

Theobromine is a methylxanthine found in cacao beans which can inhibit adenosine receptor A1 (AR1) signaling.

Specifications

Chemical Name or Material Theobromine
CAS 83-67-0
Color White
Purity Grade Notes Research
Molecular Formula C7H8N4O2
Quantity 100 mg
Synonym 3,7-Dimethylxanthine
Solubility Information DMSO : 1.1 mg/mL (6.11 mM; Need ultrasonic) ∣H2O : 1.1 mg/mL (6.11 mM; Need ultrasonic)
SMILES O=C(N1)N(C)C2=C(N(C)C=N2)C1=O
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.