missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Theobromine-d6, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Theobromine-d6 |
| CAS | 117490-40-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 2H6 H2 N4 O2 |
| Quantity | 1 mg |
| Synonym | Theophylline-Ethylenediamine EP Impurity G-d6 (Caffeine EP Impurity D-d6','Theobromine-d6) |
| InChI Formula | InChI=1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)9-7(13)11(5)2/h3H,1-2H3,(H,9,12,13)/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])N1C(=O)NC(=O)c2c1ncn2C([2H])([2H])[2H] |
| IUPAC Name | 3,7-bis(trideuteriomethyl)purine-2,6-dione |
| Molecular Weight (g/mol) | 186.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction