missing translation for 'onlineSavingsMsg'
Learn More

Theobromine, 99%

Product Code. 11493930
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493930 100 g 100g
11453824 50 g 50g
11463824 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493930 Supplier Thermo Scientific Alfa Aesar Supplier No. A11861.22
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phosphodiesterase inhibitor | CAS: 83-67-0 | C7H8N4O2 | 180.167 g/mol

Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.

Solubility
Soluble in hot methanol and dimethyl sulfoxide.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-67-0
Molecular Formula C7H8N4O2
Molecular Weight (g/mol) 180.167
MDL Number MFCD00022830
InChI Key YAPQBXQYLJRXSA-UHFFFAOYSA-N
Synonym theobromine, 3,7-dimethylxanthine, diurobromine, teobromin, theosalvose, theostene, santheose, thesodate, thesal, theobromin
PubChem CID 5429
ChEBI CHEBI:28946
IUPAC Name 3,7-dimethylpurine-2,6-dione
SMILES CN1C=NC2=C1C(=O)NC(=O)N2C

Specifications

Melting Point 345°C to 350°C
Density 1.5
Quantity 100 g
Beilstein 16464
Merck Index 14,9282
Solubility Information Soluble in hot methanol and dimethyl sulfoxide.
Formula Weight 180.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Theobromine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.