missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Themis Antibody (719945), mFluor Violet 500 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6816MFV500
This item is not returnable.
View return policy
Description
Themis Monoclonal antibody specifically detects Themis in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| Themis | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 387357 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, CyTOF | |
| 719945 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
| E. coli-derived recombinant mouse Themis, Lys122-Lys237, Accession # Q8BGW0 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction