missing translation for 'onlineSavingsMsg'
Learn More

THEM2 Antibody, DyLight 405 , Novus Biologicals™

Product Code. 18433049 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433049 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433049 Supplier Novus Biologicals Supplier No. NBP297056V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

THEM2 Polyclonal antibody specifically detects THEM2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen THEM2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias acyl-CoA thioesterase 13PNAS-27, EC 3.1.2.-, HT012, hypothalamus protein HT012, MGC4961, THEM2, Thioesterase superfamily member 2acyl-coenzyme A thioesterase 13
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human THEM2 (NP_060943.1; Met1-Asn140).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55856
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.