missing translation for 'onlineSavingsMsg'
Learn More

THEGL Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA564358

Additional Details : Weight : 0.02000kg

Product Code. 15742122

  • £427.00 / 100µL
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: IALAKSKSVH QDYLPDRDAH WPVSYATTHS KASPRIQELA NPNKRAPVRI VYYDPDVFKT KPAALKAQCS QRIWELSQ Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.

THEGL Polyclonal antibody specifically detects THEGL in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

THEGL
Polyclonal
Unconjugated
THEGL
testicular haploid expressed gene protein-like; theg spermatid protein-like; theg spermatid-like protein; THEGL
Rabbit
Antigen affinity chromatography
RUO
100506564
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P0DJG4
THEGL
Recombinant protein corresponding to Human THEGL
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only