missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THEG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570354
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 92%; Rat: 93%.
THEG gene ontology annotations related to this gene include spermatogenesis.
Specifications
| THEG | |
| Polyclonal | |
| Unconjugated | |
| THEG | |
| Cancer/testis antigen 56; CT56; testicular haploid expressed gene; testicular haploid expressed gene product; testicular haploid expressed gene protein; testis-specific; Theg; Theg homolog; theg spermatid protein; THEG1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51298 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9P2T0 | |
| THEG | |
| synthetic peptide directed towards the middle region of human THEG. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction