missing translation for 'onlineSavingsMsg'
Learn More

Theaflavin, MedChemExpress

Product Code. 16420386
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16420386

Brand: Medchem Express HYN0243

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Theaflavin is a suitable natural inhibitor against influenza A (H1N1) neuraminidase.

Theaflavin is a suitable natural inhibitor against influenza A (H1N1) neuraminidase.

Specifications

Chemical Name or Material Theaflavin
CAS 4670-05-7
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C29H24O12
Quantity 2 mg
Solubility Information DMSO : 125 mg/mL (221.44 mM; Need ultrasonic) ∣H2O : 2 mg/mL (3.54 mM; Need ultrasonic and warming)
SMILES O=C1C(O)=CC([C@@H]2[C@H](O)CC3=C(O)C=C(O)C=C3O2)=CC4=C([C@@H]5[C@H](O)CC6=C(O)C=C(O)C=C6O5)C=C(O)C(O)=C41
Molecular Weight (g/mol) 564.49
Formula Weight 564.49
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.