missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ THDOC
GREENER_CHOICE

Product Code. 18709426
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18709426

Brand: Tocris Bioscience™ 6334/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive modulator of GABAA receptors; endogenous neurosteroid

THDOC is a positive modulator of GABAA receptors. Potentiates neuronal response to low concentrations of GABA at α4β1δ GABAA receptors in vitro. Endogenous neurosteroid.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3α,5α)-3,21-Dihydroxypregnan-20-one
CAS 567-02-2
Quantity 10 mg
Molecular Formula C21H34O3
Formula Weight 334.49
Product Type Positive Allosteric Modulator
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.