missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THAP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551819
This item is not returnable.
View return policy
Description
Immunogen sequence: GISFHRLPKK DNPRRGLWLA NCQRLDPSGQ GLWDPASEYI YFCSKHFEED CFELVGISGY HRLKEGAVPT IFESFSKLRR TTKTKGHSYP PGPPEVSRLR RCRKRCSEGR GPTTPFSPPP PADVTCFP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
Chromatin-associated, histone tail-binding protein that represses transcription via recruitment of HDAC3 and nuclear hormone receptor corepressors.
Specifications
| THAP7 | |
| Polyclonal | |
| Unconjugated | |
| THAP7 | |
| THAP domain containing 7; THAP domain-containing protein 7; THAP7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80764 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BT49 | |
| THAP7 | |
| Recombinant protein corresponding to Human THAP7. Recombinant protein control fragment (Product #RP-91526). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction