missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THAP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565661
This item is not returnable.
View return policy
Description
Immunogen sequence: EDNQGKDPSKK KSQKKNLEDE KEVCPKAKSE ESFVLNETKK NIVNTDVPHQ HPELLHSSSL VKPPAPKTGS I Highest antigen sequence identity to the following orthologs - mouse 28%, rat 31%.
Has sequence-specific DNA-binding activity and can function as transcriptional repressor (in vitro). May be a regulator of cell cycle: THAP5 overexpression in human cell lines causes cell cycle arrest at G2/M phase.
Specifications
| THAP5 | |
| Polyclonal | |
| Unconjugated | |
| THAP5 | |
| THAP domain containing 5; THAP domain-containing protein 5; THAP5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 168451 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z6K1 | |
| THAP5 | |
| Recombinant Human THAP5. Recombinant protein control fragment (Product #RP-104923). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction