missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THAP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101928
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total THAP4.
THAP4 gene ontology annotations related to this gene include DNA binding; DNA-binding transcription factor activity, RNA polymerase II-specific; heme binding; metal ion binding; protein homodimerization activity.
Specifications
| THAP4 | |
| Polyclonal | |
| Unconjugated | |
| THAP4 | |
| 2010320B01Rik; CGI-36; PP238; THAP domain containing 4; THAP domain-containing protein 4; Thap4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 363291, 51078, 67026 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q642B6, Q6P3Z3, Q8WY91 | |
| THAP4 | |
| A synthesized peptide derived from human THAP4(Accession Q8WY91), corresponding to amino acid residues I342-E392. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction