missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
THAP11 Polyclonal antibody specifically detects THAP11 in Human,Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | THAP11 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CTG-B43a, CTG-B45d, HRIHFB2206, RONIN, THAP domain containing 11, THAP domain-containing protein 11 |
| Host Species | Sheep |
| Immunogen | E. coli-derived recombinant human THAP11, Gly226-Met314, Accession # Q96EK4 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?