missing translation for 'onlineSavingsMsg'
Learn More

THAP11 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18401649 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401649 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401649 Supplier Novus Biologicals Supplier No. NBP297155AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

THAP11 Polyclonal antibody specifically detects THAP11 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen THAP11
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias CTG-B43a, CTG-B45d, HRIHFB2206, RONIN, THAP domain containing 11, THAP domain-containing protein 11
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human THAP11 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 57215
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.