missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THAP1 Monoclonal Antibody (8D10C8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531931
This item is not returnable.
View return policy
Description
MA5-31931 has been tested in indirect ELISA.
DNA-binding transcription regulator that regulates endothelial cell proliferation and G1/S cell-cycle progression. Specifically binds the 5'-[AT]NTNN[GT]GGCA[AGT]-3' core DNA sequence and acts by modulating expression of pRB-E2F cell-cycle target genes, including RRM1. Component of a THAP1/THAP3-HCFC1-OGT complex that is required for the regulation of the transcriptional activity of RRM1. May also have pro-apoptopic activity by potentiating both serum-withdrawal and TNF-induced apoptosis.
Specifications
| THAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9NVV9 | |
| Thap1 | |
| Purified recombinant fragment of human THAP1 (AA: 1-213) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 8D10C8 | |
| Unconjugated | |
| Thap1 | |
| 4833431A01Rik; AW490810; DYT6; nuclear proapoptotic factor; THAP domain containing 1; THAP domain containing, apoptosis associated protein 1; THAP domain protein 1; THAP domain-containing apoptosis-associated protein 1; THAP domain-containing protein 1; THAP1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 55145 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction