missing translation for 'onlineSavingsMsg'
Learn More

Thallium Isotopic Standard: Tl @ 10 μg/mL in 2% HNO3 (Enriched in Tl-203), VHG™

Product Code. 30003147
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003147 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003147 Supplier VHG Labs Supplier No. VHGLIS203TL50
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Thallium Isotopic Standard: Tl @ 10 μg/mL in 2% HNO3 (Enriched in Tl-203)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material total elem
Methods ICP-MS
Analyte or Component Names total elem
Shipping Condition Room Temperature
IUPAC Name total elem
Solvent or Matrix Dilute HNO3
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.