missing translation for 'onlineSavingsMsg'
Learn More

Thallium(III) trifluoroacetate, 95%

Product Code. 11369129
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11369129

Brand: Thermo Scientific Alfa Aesar 087913.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23586-53-0 | C6F9O6Tl | 543.426 g/mol

Thallium(III) trifluoroacetate is an oxidizing agent used in the synthesis of S-substituted cysteine peptides and proteins containing disulfide bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23586-53-0
Molecular Formula C6F9O6Tl
Molecular Weight (g/mol) 543.426
MDL Number MFCD00000414
InChI Key PSHNNUKOUQCMSG-UHFFFAOYSA-K
Synonym thallium iii trifluoroacetate, acetic acid, trifluoro-, thallium 3+ salt, acetic acid,2,2,2-trifluoro-, thallium 3+ salt 3:1, thallium 3+ ; 2,2,2-trifluoroacetate, thallium tris trifluoroacetate, trifluoroacetic acid thallium iii, thallic trifluoroacetate, tech, thallium 3+ tritrifluoroacetate
PubChem CID 90200
IUPAC Name thallium(3+);2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Tl+3]

Specifications

Melting Point 213°C (decomposition)
Physical Form Solid
Quantity 50 g
UN Number UN1707
Solubility Information Soluble in water (decomposes).
Formula Weight 543.42
Percent Purity 95%
Sensitivity Hygroscopic
Chemical Name or Material Thallium (III) trifluoroacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.