missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallium(I) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Thallium(I) iodide is used in mercury lamp to enhance its performance. It is mainly used for underwater lighting, since the blue green light produced has least absorption in water. Scintillator made by using iodide salt of thallium, sodium and cesium is utilized for radiation scintillation detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 824°C |
Melting Point | 440°C |
Color | Yellow |
Physical Form | Beads |
Quantity | 250 g |
Assay Percent Range | 99.999% metal basis |
Solubility Information | Insoluble in water. |
Formula Weight | 331.27 |
Odor | Odorless |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'