missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) fluoride, 97%

Product Code. 11374899
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374899 100 g 100g
11384899 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374899 Supplier Thermo Scientific Alfa Aesar Supplier No. 078111.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-27-7 | FTl | 223.38 g/mol

Thallium(I) fluoride is used as chemical reagents, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-27-7
Molecular Formula FTl
Molecular Weight (g/mol) 223.38
MDL Number MFCD00049607
InChI Key CULOEOTWMUCRSJ-UHFFFAOYSA-M
Synonym thallium i fluoride, thallium fluoride, thallium monofluoride, thallium fluoride ic
PubChem CID 62675
SMILES [F-].[Tl+]

Specifications

Boiling Point 655°C
Physical Form Powder
Quantity 100 g
UN Number UN1707
Merck Index 14,9262
Solubility Information Very soluble in water
IUPAC Name fluorothallium
Formula Weight 223.37
Percent Purity 97%
Density 8.36 g/mL
Chemical Name or Material Thallium (I) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.