missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) ethoxide, 95%, Thermo Scientific Chemicals

Product Code. 11374909
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374909 25 g 25g
11364909 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374909 Supplier Thermo Scientific Alfa Aesar Supplier No. 078131.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20398-06-5
Molecular Formula C2H5OTl
Molecular Weight (g/mol) 249.441
MDL Number MFCD00009072
InChI Key DZFYOYRNBGNPJW-UHFFFAOYSA-N
Synonym thallous ethoxide, thallium i ethanolate, ethoxythallium i, ethanol, thallium 1+ salt, ethyl alcohol, thallium i, ethanol, thallium 1+ salt 1:1, thallium ethoxide, ethanolate; thallium 1+, thallium i ethoxide 5g
PubChem CID 30143
IUPAC Name ethanolate;thallium(1+)
SMILES CC[O-].[Tl+]

Specifications

Boiling Point 130°C (decomposition)
Melting Point -3°C
Physical Form Liquid
Quantity 25 g
Linear Formula TlOCH2CH3
UN Number UN2810
Flash Point >110°C (230°F)
Formula Weight 249.43
Percent Purity 98%
Refractive Index 1.6714
Sensitivity Moisture Sensitive
Density 3.493g/mL
Chemical Name or Material Thallium (I) ethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.