missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) chloride, ultra dry, 99.999% (metals basis)

Product Code. 11328577
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11328577

Brand: Thermo Scientific Alfa Aesar 035721.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7791-12-0 | ClTl | 239.83 g/mol

It is used as a catalyst in chlorination's and in suntan lamps. The low solubility of TlCl is exploited in chemical synthesis: treatment of metal chloride complexes with TlPF6, gives the corresponding metal hexafluorophosphate derivative. It is used as a tracer in medical imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7791-12-0
Molecular Formula ClTl
Molecular Weight (g/mol) 239.83
MDL Number MFCD00011274
InChI Key GBECUEIQVRDUKB-UHFFFAOYSA-M
Synonym thallium chloride, thallium i chloride, thallous chloride, thallium monochloride, thallium 1+ chloride, tlcl, thallium chloride tlcl, thallium chloride van, rcra waste number u216, rcra waste no. u216
PubChem CID 24642
ChEBI CHEBI:37117
IUPAC Name chlorothallium
SMILES Cl[Tl]

Specifikationer

Melting Point 430°C
Physical Form Beads
Quantity 25 g
UN Number UN1707
Merck Index 14,9260
Solubility Information Slightly soluble in water. Insoluble in alcohol,and acetone.
Formula Weight 239.82
Percent Purity 99.999%
Packaging Ampule
Refractive Index 2.247
Sensitivity Air Sensitive
Density 7.0 g/mL
Assay (metals basis)
Chemical Name or Material Thallium(I) chloride, Ultra dry
Visa mer Visa mindre

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.