missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) chloride, 99%

Product Code. 10092730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10092730 100 g Glass bottle 100g
10308540 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10092730 Supplier Thermo Scientific Acros Supplier No. 208881000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7791-12-0 | ClTl | 239.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-12-0
Molecular Formula ClTl
Molecular Weight (g/mol) 239.82
MDL Number MFCD00011274
InChI Key GBECUEIQVRDUKB-UHFFFAOYSA-M
Synonym thallium chloride, thallium i chloride, thallous chloride, thallium monochloride, thallium 1+ chloride, tlcl, thallium chloride tlcl, thallium chloride van, rcra waste number u216, rcra waste no. u216
PubChem CID 24642
ChEBI CHEBI:37117
IUPAC Name chlorothallium
SMILES Cl[Tl]

Specifications

Boiling Point 720°C
Melting Point 430°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 98.5% min
Linear Formula TICl
Merck Index 15, 9409
Solubility Information Solubility in water: 0.38%. Other solubilities: soluble in 70 parts boiling water,insoluble in alcohol and ammonium hydroxide,hcl decreases its solubulity in water
Formula Weight 239.82
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 7
Density 7.0000g/mL
Chemical Name or Material Thallium(I) chloride, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thallium(I) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.