missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

£105.00 - £364.00

Chemical Identifiers

CAS 7789-40-4
MDL Number MFCD00011273
Synonym Thallous bromide
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398567
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035720.09
10 g
£105.00
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11308577
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035720.18
50 g
£364.00
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thallium(I) bromide is used in blue sensitive photodetectors. It is also used as a real-time x-ray image sensor. Further, it acts as a standard for elemental thallium. In addition to this, it is a compound semiconductor used in room temperature X- and gamma-ray detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

7789-40-4
MFCD00011273
Thallous bromide

Specifications

∼460°C
Beads
UN1707
Thallous bromide
284.28
Ampule
7.5 g/mL
Thallium(I) bromide, Ultra dry
7789-40-4
MFCD00011273
14,9258
Insoluble in water.
99.998%
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

missing translation for 'einecsNumber' : 232-163-0

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only