missing translation for 'onlineSavingsMsg'
Learn More

Thallium AA Standard: Tl @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17998689
Click to view available options
Quantity:
100 mL
500 mL
missing translation for 'unitSize'
100mL
500mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 17998689

missing translation for 'mfr': VHG Labs VHGAATLN100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Thallium AA Standard: Tl @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thallium
CAS 7440-28-0
Methods AA/GFAA
Analyte or Component Names Thallium
Shipping Condition Room Temperature
Molecular Formula Tl
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Ramor, Thallium, metallic, Thallium, elemental
InChI Formula InChI=1S/Tl
SMILES [Tl]
IUPAC Name thallium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 204.38
Quantity 100 mL
Formula Weight 204.974 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.