missing translation for 'onlineSavingsMsg'
Learn More

Thallic acetate, 98%, Thermo Scientific™

Product Code. 10762084 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10762084

Brand: Thermo Scientific Acros 420750050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2570-63-0
Molecular Formula C6H9O6Tl
Molecular Weight (g/mol) 381.51
MDL Number MFCD00008698
Synonym Acetic acid, thallium(III) salt

Specifications

Melting Point 182°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 98%
Linear Formula (CH3CO2)3Tl
Beilstein 02, 96
Fieser 01,1150; 02,406; 03,286; 04,492; 05,655; 07,360
Merck Index 15, 9406
Solubility Information Solubility in water: decomposes
Formula Weight 381.51
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Thallic acetate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.