missing translation for 'onlineSavingsMsg'
Learn More

Thallic acetate, 98%, Thermo Scientific™

Product Code. 10299460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10299460 100 g Glass bottle 100g
10762084 5 g Glass bottle 5g
10381781 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10299460 Supplier Thermo Scientific Acros Supplier No. 420751000
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2570-63-0
Molecular Formula C6H9O6Tl
Molecular Weight (g/mol) 381.51
MDL Number MFCD00008698
Synonym Acetic acid, thallium(III) salt

Specifications

Melting Point 182°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 98%
Linear Formula (CH3CO2)3Tl
Beilstein 02, 96
Fieser 01,1150; 02,406; 03,286; 04,492; 05,655; 07,360
Merck Index 15, 9406
Solubility Information Solubility in water: decomposes
Formula Weight 381.51
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Thallic acetate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.