missing translation for 'onlineSavingsMsg'
Learn More

Thalidomide-O-COOH, MedChemExpress

Product Code. 16451556
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451556 500 mg 500mg
16470086 2 g 2g
16411009 1 g 1g
16404906 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16451556 Supplier Medchem Express Supplier No. HY103597
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Thalidomide-O-COOH (Cereblon ligand 3) is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide-O-COOH (Cereblon ligand 3) can be connected to the ligand for protein by a linker to form PROTACs.

Thalidomide-O-COOH (Cereblon ligand 3) is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide-O-COOH (Cereblon ligand 3) can be connected to the ligand for protein by a linker to form PROTACs.

Specifications

Chemical Name or Material Thalidomide-O-COOH
CAS 1061605-21-7
Color White
Purity Grade Notes Research
Molecular Formula C15H12N2O7
Quantity 500 mg
Synonym Cereblon ligand 3 E3 ligase Ligand 3
Solubility Information DMSO : ≥ 42 mg/mL (126.41 mM)
SMILES O=C(N(C(CCC1=O)C(N1)=O)C2=O)C(C2=CC=C3)=C3OCC(O)=O
Molecular Weight (g/mol) 332.26
Formula Weight 332.26
Percent Purity 96.75%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.