missing translation for 'onlineSavingsMsg'
Learn More

Thalidomide-O-amido-C4-NH2 TFA, MedChemExpress

Product Code. 16491019
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
This item is not returnable. View return policy

Product Code. 16491019

Brand: Medchem Express HY103613

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Thalidomide-O-amido-C4-NH2 TFA (Cereblon Ligand-Linker Conjugates 6 TFA) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.

Thalidomide-O-amido-C4-NH2 TFA (Cereblon Ligand-Linker Conjugates 6 TFA) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.

Specifications

Chemical Name or Material Thalidomide-O-amido-C4-NH2 TFA
CAS 1799711-25-3
Color White
Purity Grade Notes Research
Molecular Formula C21H23F3N4O8
Quantity 1 g
Synonym Cereblon Ligand-Linker Conjugates 6 TFAE3 ligase Ligand-Linker Conjugates 19 TFA
Solubility Information DMSO : 42.86 mg/mL (82.99 mM; Need ultrasonic)
SMILES NCCCCNC(COC1=C(C(N2C(CCC3=O)C(N3)=O)=O)C(C2=O)=CC=C1)=O.FC(F)(F)C(O)=O
Molecular Weight (g/mol) 516.42
Formula Weight 516.42
Percent Purity 96.6%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.