missing translation for 'onlineSavingsMsg'
Learn More

S1P1 agonist III, MedChemExpress

Product Code. 16411577
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16411577

Brand: Medchem Express HY12835

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress S1P1 Agonist III is a potent and orally active S1P1 agonist with EC50 of 18 nM; no activity on S1P3.

S1P1 Agonist III is a potent and orally active S1P1 agonist with EC50 of 18 nM

Specifications

Chemical Name or Material S1P1 agonist III
CAS 1324003-64-6
Color White
Purity Grade Notes Research
Molecular Formula C21H16F3N3O3
Quantity 50 mg
Solubility Information DMSO : 62.5 mg/mL (150.47 mM; Need ultrasonic)
SMILES COC1=CC=NC=C1C(NC(NC2=CC(C(F)(F)F)=C(C3=CC=CC=C3)C=C2)=O)=O
Molecular Weight (g/mol) 415.37
Formula Weight 415.37
Percent Purity 97.14%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.