missing translation for 'onlineSavingsMsg'
Learn More

Thalidomide, MP Biomedicals™

Product Code. 11489941 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
50 g
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489941 100 mg 100mg
11479941 50 g 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489941 Supplier MP Biomedicals Supplier No. 0215875380
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 50-35-1
Molecular Formula C13H10N2O4
Molecular Weight (g/mol) 258.23
MDL Number MFCD00153873
InChI Key UEJJHQNACJXSKW-UHFFFAOYNA-N
Synonym thalidomide, thalomid, +/--thalidomide, sedoval, 2-2,6-dioxopiperidin-3-yl isoindoline-1,3-dione, corronarobetin, psycholiquid, psychotablets, theophilcholine, algosediv
PubChem CID 5426
ChEBI CHEBI:74947
IUPAC Name 2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1N(C2CCC(=O)NC2=O)C(=O)C2=CC=CC=C12

Specifications

Color White
Quantity 100 mg
Physical Form Solid
Chemical Name or Material Thalidomide
compliance-icons compliance-icons
Product Identifier
  • thalidomide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
Hazard Statement
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: thalidomide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.