missing translation for 'onlineSavingsMsg'
Learn More

Thalidomide 4'-oxyacetamide-PEG4-amine
GREENER_CHOICE

Product Code. 18763998
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 18763998

Brand: Tocris Bioscience 6468/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PROTACs (Proteolysis Targeting Chimeras)

Thalidomide 4'-oxyacetamide-PEG4-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

This product has been recently renamed. The previous name for this product was Thalidomide - linker 2

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material PROTAC Toolbox
CAS 2245697-85-0
Synonym Thalidomide - linker 2
Quantity 25 mg
Molecular Formula C25H34N4O10.HCl
Formula Weight 587.02
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.