missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Thalidomide-4-hydroxyacetate
GREENER_CHOICE

Product Code. 18729768
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18729768 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18729768 Supplier Tocris Bioscience™ Supplier No. 6466/100
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Carboxylic acid-functionalized Thalidomide; cereblon ligand for PROTACs

Thalidomide-4-hydroxyacetate is a derivative of Thalidomide (Cat.No. 0652), commonly used as a precursor to PROTAC® that hijacks cereblon as the E3 ubiquitin ligase component. Supplied with a carboxylic acid functional handle at a position known not to significantly affect binding to cereblon, for ready conjugation to a linker/target protein ligand.

Thalidomide-4-hydroxyacetate has been recently renamed. The previous name for this product was TC E3 5031.

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

Require a different derivative? Get in touch .

TRUSTED_SUSTAINABILITY

Specifications

CAS 1061605-21-7
Synonym 2-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]oxy]acetic acid
Quantity 100 mg
Target Degrader Building Blocks
Molecular Formula C15H12N2O7
Formula Weight 332.26
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.