missing translation for 'onlineSavingsMsg'
Learn More

Th-POK Mouse anti-Human, Clone: PCRP-ZBTB7B-1F7, Novus Biologicals™

Product Code. 18655967 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18655967 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18655967 Supplier Novus Biologicals Supplier No. NBP307758100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Th-POK Monoclonal specifically detects Th-POK in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Th-POK
Applications Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
Classification Monoclonal
Clone PCRP-ZBTB7B-1F7
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias hcKroxc-Krox, Krueppel-related zinc finger protein cKrox, kruppel-related zinc finger protein hcKrox, T-helper-inducing POZ/Krueppel-like factor, THPOK, ZBTB15zinc finger protein 67 homolog (mouse), zfp-67, ZFP67, zinc finger and BTB domain containing 15, zinc finger and BTB domain containing 7B, Zinc finger and BTB domain-containing protein 15, zinc finger and BTB domain-containing protein 7B, Zinc finger protein 67 homolog, Zinc finger protein 857B, Zinc finger protein Th-POK, ZNF857BDKFZp686G01254
Host Species Mouse
Immunogen Recombinant full-length human Th-POK protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51043
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.