missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Th-POK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88078
This item is not returnable.
View return policy
Description
Th-POK Polyclonal specifically detects Th-POK in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Th-POK | |
| Polyclonal | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| hcKroxc-Krox, Krueppel-related zinc finger protein cKrox, kruppel-related zinc finger protein hcKrox, T-helper-inducing POZ/Krueppel-like factor, THPOK, ZBTB15zinc finger protein 67 homolog (mouse), zfp-67, ZFP67, zinc finger and BTB domain containing 15, zinc finger and BTB domain containing 7B, Zinc finger and BTB domain-containing protein 15, zinc finger and BTB domain-containing protein 7B, Zinc finger protein 67 homolog, Zinc finger protein 857B, Zinc finger protein Th-POK, ZNF857BDKFZp686G01254 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 51043 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ZBTB7B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LLEFAYTATLTTSSANMPAVLQAARLLEIPCVIAACMEILQGSGLEAPSPDEDDCERARQYLEAFATATASGVPNGEDSPPQVPLPPPPPPPPRPVARRSRKPRKAFLQTKG | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction