missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ TH 5487

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TH 5487 is a selective 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor (IC50 = 342 nM). Selective for OGG1 over other base excision repair enzymes. Inhibits OGG1 binding to 8-oxoG in DNA and suppresses proinflammatory gene expression in TNF-α stimulated lung epithelial cells in vitro. Also inhibits proinflamatory gene expression and neutrophil recruitment in TNF-α-challenged mouse lungs in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Molecular Formula | C19H18BrIN4O2 |
| Formula Weight | 541.18 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction