missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ TH 1760
Description
TH 1760 is a potent and selective NUDT15 (nudix hydrolase 15) inhibitor (IC50 = 29 nM). Displays selectivity for NUDT15 when assayed at 100 μM against other Nudix enzymes (MTH1, NUDT2, 5, 9, 12, 14, 18, 22) and pyrophosphatases or at 10 μM when assayed with a Eurofins Cerep Safety Screen44 panel. In vitro, TH 1760 treatment sensitizes cancer cell lines (NB4 and HL-60) to thiopurine.
Specifications
Specifications
| Chemical Name or Material | 6-[[4-(1 H-Indol-5-ylcarbonyl)-1-piperazinyl]sulfonyl]-2(3 H)-benzoxazolone |
| CAS | 2567914-01-4 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | Hydrolase Inhibitors |
| Molecular Formula | C20H18N4O5S |
| Purity | 0.95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?