missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TGX 221
Description
TGX 221 is a potent and selective PI 3-kinase β inhibitor (IC50 values are 0.007, 0.1, 3.5 and 5 μM for the β, δ, γ and α isoforms, respectively). Shows >1,000-fold selectively for PI 3-kinase β over a range of other kinases. Inhibits thrombus formation in animal models.
Specifications
Specifications
| Chemical Name or Material | 7-Methyl-2-(4-morpholinyl)-9-[1-(phenylamino)ethyl]-4H-pyrido[1,2-a]pyrimidin-4-one |
| CAS | 663619-89-4 |
| Quantity | 10 mg |
| Target | PI 3-Kinase Inhibitors |
| Molecular Formula | C21H24N4O2 |
| Formula Weight | 364.44 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?