missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13034-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a Ro RNP-binding protein. It interacts with Ro RNPs and their interaction is thought to represent a gain of function for Ro RNPs. This protein also forms a ternary complex with far upstream element (FUSE) and FUSE-binding protein. It can repress a c-myc reporter via the FUSE. It is also known to target transcription factor IIH and inhibit activated transcription. This gene is implicated in the xeroderma pigmentosum disorder. There are two alternatively spliced transcript variants of this gene encoding different isoforms. There seems to be evidence of multiple polyadenylation sites for this gene.Specifications
| TGS1 | |
| Polyclonal | |
| Unconjugated | |
| TGS1 | |
| CLL associated antigen KW 2, HCA137, NCOA6IP, PIMT, PIPMT, TGS1, Trimethylguanosine synthase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116940, 312947, 96764 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P85107, Q923W1, Q96RS0 | |
| TGS1 | |
| TGS1 Fusion Protein Ag3683 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction