missing translation for 'onlineSavingsMsg'
Learn More

TGR5 Receptor Agonist, MedChemExpress

Product Code. 16448617
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16448617

Brand: Medchem Express HY14229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TGR5 Receptor Agonist (CCDC), a potent Takeda G protein-coupled receptor 5 (TGR5; GPCR19) agonist, shows improved potency in the U2-OS cells and melanophore cells with pEC50s of 6.8 and 7.5, respectively.

TGR5 Receptor Agonist can induce peripheral and central hypersensitivity to bladder distension in mice, and increase intracellular Ca2+ concentration. TGR5 Receptor Agonist can also reduces food intake and improves insulin responsiveness, in diet-induced obese mice. TGR5 Receptor Agonist can be used to research diabetes, bladder hypersensitivity and anti-obesity.

Specifications

Chemical Name or Material TGR5 Receptor Agonist
Physical Form Solid
CAS 1197300-24-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C18H14Cl2N2O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym CCDC
Solubility Information DMSO : ≥ 48 mg/mL (132.88 mM)
SMILES ClC1=CC=CC=C1C2=NOC(C)=C2C(N(C)C3=CC=C(Cl)C=C3)=O
Molecular Weight (g/mol) 361.22
Formula Weight 361.22
Percent Purity 99.86%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.