missing translation for 'onlineSavingsMsg'
Learn More

TGR5/GPBAR1 Antibody, PE, Novus Biologicals™

Product Code. 18421692
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421692 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421692 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP223669PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TGR5/GPBAR1 Polyclonal antibody specifically detects TGR5/GPBAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGR5/GPBAR1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation PBS
Gene Alias BG37hBG37, G protein-coupled bile acid receptor 1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR
Host Species Rabbit
Immunogen Synthetic peptide from C-terminus of the human TGR5/GPBAR1 protein [UniProt Q8TDU6]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 151306
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.