missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGR5/GPBAR1 Antibody (409522) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4286JF585
This item is not returnable.
View return policy
Description
TGR5/GPBAR1 Monoclonal antibody specifically detects TGR5/GPBAR1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| TGR5/GPBAR1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| BG37hBG37, G protein-coupled bile acid receptor 1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR | |
| NS0 mouse myeloma cell line transfected with human TGR5/GPBAR1, Met1-Asn330, Accession # Q8TDU6 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 409522 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 151306 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction