missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGN46 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13573-1-AP-150UL
This item is not returnable.
View return policy
Description
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.Specifications
| TGN46 | |
| Polyclonal | |
| Unconjugated | |
| TGOLN2 | |
| Tgn38, TGN38 homolog, TGN46, TGN48, TGN51, TGOLN2, trans Golgi network protein 2, TTGN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10618 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43493 | |
| TGOLN2 | |
| TGN46 Fusion Protein Ag4470 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction