missing translation for 'onlineSavingsMsg'
Learn More

TGN46 Antibody, Novus Biologicals™

Product Code. 18424111 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424111 25 μL 25µL
18302770 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18424111 Supplier Novus Biologicals Supplier No. NBP18694825ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TGN46 Polyclonal specifically detects TGN46 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGN46
Applications Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias TGN 46, TGN 48, TGN 51, TGN38, TGN-38, TGN38 homolog, TGN46, TGN-46, TGN48, TGN51, TGOLN 2, TGOLN2, trans golgi network 38, trans golgi network 46, Trans Golgi network integral membrane protein 2, Trans Golgi network integral membrane protein 2 [Precursor], Trans Golgi network protein (46 48 51kD isoforms), Trans golgi network protein 2, Trans Golgi network protein TGN51, Trans-Golgi network integral membrane protein 2, Trans-Golgi network protein 2, Trans-Golgi network protein TGN51, TTGN 2, TTGN2
Gene Symbols TGOLN2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:KDVPNKSGAEKQTPKDGSNKSGAEEQGPIDGPSKSGAEEQTSKDSPNKVVPEQPSWKDHSKPISNPSDNKELPKADTNQLADKGKLSPHAFKTESGEETDLISPPQEEVKSSEPTEDVEPKEAEDDDTGPEEGSPPKEEKEKMSGSAS
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Trans golgi Network Markers
Primary or Secondary Primary
Gene ID (Entrez) 10618
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.