missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGM7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27748-1-AP-20UL
This item is not returnable.
View return policy
Description
Transglutaminases (TGM; EC 2.3.2.13) are a family of structurally and functionally related enzymes that stabilize protein assemblies through the formation of gamma-glutamyl-epsilon lysine crosslinks. For additional background information on transglutaminases, see TGM1 (MIM 190195).Specifications
| TGM7 | |
| Polyclonal | |
| Unconjugated | |
| TGM7 | |
| TG(Z), TGase 7, TGase Z, TGM7, TGMZ, TGZ, transglutaminase 7, Transglutaminase Z | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 102552199, 116179, 640543 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96PF1 | |
| Tgm7 | |
| TGM7 Fusion Protein Ag26875 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction