missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGM5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577108
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the transglutaminase family. The encoded protein catalyzes formation of protein cross-links between glutamine and lysine residues, often resulting in stabilization of protein assemblies. This reaction is calcium dependent. Mutations in this gene have been associated with acral peeling skin syndrome.
Specifications
| TGM5 | |
| Polyclonal | |
| Unconjugated | |
| Tgm5 | |
| 2310007C07Rik; MGC141907; protein-glutamine gamma-glutamyltransferase 5; PSS2; TG(X); TGase 5; TGase X; TGASE5; TGase-5; TGASEX; Tgm5; TGM6; TGMX; TGX; transglutaminase 5; transglutaminase V; transglutaminase X; Transglutaminase-5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 691929, 74176, 9333 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43548, Q9D7I9 | |
| Tgm5 | |
| Recombinant full length Human TGM5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction