missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148264
This item is not returnable.
View return policy
Description
TGM3 Polyclonal Antibody for IHC (P)
Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. The protein encoded by this gene consists of two polypeptide chains activated from a single precursor protein by proteolysis. The encoded protein is involved the later stages of cell envelope formation in the epidermis and hair follicle.
Specifications
| TGM3 | |
| Polyclonal | |
| Unconjugated | |
| TGM3 | |
| AI893889; E polypeptide; E polypeptide, protein-glutamine-gamma-glutamyltransferase; MGC126249; MGC126250; protein-glutamine gamma-glutamyltransferase E; Protein-glutamine gamma-glutamyltransferase E 27 kDa non-catalytic chain; Protein-glutamine gamma-glutamyltransferase E 50 kDa catalytic chain; protein-glutamine-gamma-glutamyltransferase; RGD1561831; TG E; TG(E); TGase E; TGase TGase-3; Tgase3; TGase-3; TGE; TGM3; transglutaminase 3; transglutaminase 3 (E polypeptide, protein-glutamine-gamma-glutamyltransferase); transglutaminase 3, E polypeptide; transglutaminase E; Transglutaminase-3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7053 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q08188 | |
| TGM3 | |
| E. coli-derived Human Transglutaminase 3/TGM3 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction