missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TGM2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA529356

Product Code. 13493527

  • £443.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: A549. Predicted reactivity: Mouse (87%), Rat (86%), Bovine (87%), Guinea pig (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Transglutaminase (TGase) catalyses the crosslink of proteins by forming e-(g-glutamyl) lysine isopeptide bonds and requires the binding of Ca2+ for its activity. In mammals, eight distinct TGase isoenzymes have been identified. Tissue transglutaminase (tTGase), also known as TGase 2, has four distinct domains: N-terminal b-sandwich, catalytic core and two C-terminal b-barrel domains. tTGase may have a role in cell death, cell proliferation, cell differentiation, and receptor-mediated endocytosis. In the Alzheimer's disease brain, the elevated tTGase activity is manifested by polymerization of a number of proteins, including Ab peptide, b-amyloid precursor protein and the tau protein, with formation of neurofibrillary tangles.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TGM2
Polyclonal
Unconjugated
TGM2
C polypeptide; C polypeptide, protein-glutamine-gamma-glutamyltransferase; G alpha h; G[a]h; G-ALPHA-h; GNAH; Protein-glutamine gamma-glutamyltransferase 2; protein-glutamine-gamma-glutamyltransferase; RP5-1054A22.2; TG C; TG(C); TG2; TGase C; TGase H; TGase2; TGase-2; TGase-H; TgaseII; TGC; TGM2; Tissue transglutaminase; Tissue type transglutaminase; tissue-type transglutaminase; transglutaminase; transglutaminase 2; transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase); Transglutaminase 2 C polypeptide; transglutaminase 2, C polypeptide; transglutaminase C; Transglutaminase H; transglutaminase-2; tTG; tTGas
Rabbit
Antigen affinity chromatography
RUO
21817, 7052
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P21980, P21981
TGM2
Recombinant protein encompassing a sequence within the center region of human Transglutaminase 2. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.