missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TGM2 Monoclonal Antibody (CUB 7402)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA512739

Product Code. 11364703

  • £466.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-12739 targets Transglutaminase II in IF, IHC (P), IP, and WB applications and shows reactivity with Canine, Guinea Pig, Human, and Rabbit samples. The MA5-12739 immunogen is purified guinea pig liver TGase II.

Transglutaminase (TGase) catalyses the crosslink of proteins by forming e-(g-glutamyl) lysine isopeptide bonds and requires the binding of Ca2+ for its activity. In mammals, eight distinct TGase isoenzymes have been identified. Tissue transglutaminase (tTGase), also known as TGase 2, has four distinct domains: N-terminal b-sandwich, catalytic core and two C-terminal b-barrel domains. tTGase may have a role in cell death, cell proliferation, cell differentiation, and receptor-mediated endocytosis. In the Alzheimer's disease brain, the elevated tTGase activity is manifested by polymerization of a number of proteins, including Ab peptide, b-amyloid precursor protein and the tau protein, with formation of neurofibrillary tangles.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TGM2
Monoclonal
0.02 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P08587, P21980
TGM2
Purified guinea pig liver TGase II.
500 μL
Primary
Canine, Guinea Pig, Human, Rabbit
Antibody
IgG1 κ
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
CUB 7402
Unconjugated
TGM2
C polypeptide; C polypeptide, protein-glutamine-gamma-glutamyltransferase; G alpha h; G[a]h; G-ALPHA-h; GNAH; Protein-glutamine gamma-glutamyltransferase 2; protein-glutamine-gamma-glutamyltransferase; RP5-1054A22.2; TG C; TG(C); TG2; TGase C; TGase H; TGase2; TGase-2; TGase-H; TgaseII; TGC; TGM2; Tissue transglutaminase; Tissue type transglutaminase; tissue-type transglutaminase; transglutaminase; transglutaminase 2; transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase); Transglutaminase 2 C polypeptide; transglutaminase 2, C polypeptide; transglutaminase C; Transglutaminase H; transglutaminase-2; tTG; tTGas
Mouse
Protein G
RUO
100352148, 100379228, 485867, 7052
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.