missing translation for 'onlineSavingsMsg'
Learn More

TGIF1 Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30732026 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732026 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732026 Supplier Novus Biologicals Supplier No. AF7555MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

TGIF1 Polyclonal antibody specifically detects TGIF1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGIF1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias 5'-TG-3' interacting factor, homeobox protein TGIF, homeobox protein TGIF1, HPE4, MGC39747, TALE homeobox TG-interacting factor, TGFB-induced factor (TALE family homeobox), TGFB-induced factor homeobox 1, TGIFMGC5066,5'-TG-3'-interacting factor 1, transforming growth factor-beta-induced factor
Host Species Sheep
Immunogen E. coli-derived recombinant human TGIF1, Lys245-Ser359, Accession # Q15583
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7050
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.