missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGFBR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112740
This item is not returnable.
View return policy
Description
TGFBR3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications
| TGFBR3 | |
| Polyclonal | |
| Unconjugated | |
| TGFBR3 | |
| 1110036H20Rik; AU015626; AW215636; betaglycan; betaglycan proteoglycan; BGCAN; OTTHUMP00000011787; TBRIII; tgf beta receptor 3; TGF-beta receptor type 3; TGF-beta receptor type III; Tgfbr3; TGFR-3; transforming growth factor beta receptor 3; transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; transforming growth factor, beta receptor 3; transforming growth factor, beta receptor III | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7049 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q03167 | |
| TGFBR3 | |
| Recombinant Human Transforming growth factor beta receptor type 3 protein (140-340AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction